missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibutylformamide, 99+%, Thermo Scientific Chemicals

Product Code. 10214380 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
500 mL
Packaging:
Glass bottle
Unit Size:
10mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10214380 500 mL Glass bottle 500mL
10204380 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214380 Supplier Thermo Scientific Acros Supplier No. 112985000
  • ONLINE EXCLUSIVE PRICE
    675.00 €
    654.00 € / 500mL
    Save 21.00 €
    3% Off
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761-65-9
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD00003289
InChI Key NZMAJUHVSZBJHL-UHFFFAOYSA-N
Synonym dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
PubChem CID 12975
IUPAC Name N,N-dibutylformamide
SMILES CCCCN(CCCC)C=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Density 0.8600g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point 100°C
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula HCON[(CH2)3CH3]2
Refractive Index 1.4430 to 1.4450
Quantity 500 mL
Beilstein 03,443F
Specific Gravity 0.86
Formula Weight 157.26
Percent Purity 99+%
Chemical Name or Material N, N-Dibutylformamide, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.