missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylcarbodiimide, TRC

Product Code. 30375416
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30375416 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375416 Supplier Toronto Research Chemicals Supplier No. TRCD438500500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Dicyclohexylcarbodiimide
CAS 538-75-0
Purity Grade Notes GC
Molecular Formula C13 H22 N2
Quantity 500 g
Synonym 1,3-Dicyclohexylcarbodiimide, Bis(cyclohexyl)carbodiimide, DCC, DCCD, DCCI, Dicyclohexylcarbodiimide, N,N’-Methanetetraylbis[cyclohexanamine], NSC 30022, NSC 53373, NSC 57182
SMILES C1CCC(CC1)N=C=NC2CCCCC2
Molecular Weight (g/mol) 206.33
Formula Weight 206.1783
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.