missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylmorpholine-4-carboxamidine, 98+%, Thermo Scientific™

Product Code. 15472716
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472716 100 g 100g
15462716 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472716 Supplier Thermo Scientific Alfa Aesar Supplier No. H56480.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4975-73-9
Molecular Formula C17H31N3O
Molecular Weight (g/mol) 293.455
MDL Number MFCD00063252
InChI Key OZNYZQOTXQSUJM-UHFFFAOYSA-N
Synonym n,n'-dicyclohexyl-4-morpholinecarboxamidine, n,n'-dicyclohexyl-4-morpholine carboxamidine, n,n'-dicyclohexyl-1-morpholinoformamidine, n,n'-dicyclohexylmorpholine-4-carboxamidine, 4-morpholinecarboximidamide, n,n'-dicyclohexyl, formamidine, n,n'-dicyclohexyl-1-morpholino, n,n'-dicyclohexyl-4-morpholine-carboxamidine, z-n,n'-dicyclohexylmorpholine-4-carboximidamide, 4-n,n'-dicyclohexylamidino morpholine, n,n'-dicyclohexyl-4-morpholinecarboximidamide
PubChem CID 21079
IUPAC Name N,N'-dicyclohexylmorpholine-4-carboximidamide
SMILES C1CCC(CC1)NC(=NC2CCCCC2)N3CCOCC3

Specifications

Melting Point 105°C to 107°C
Quantity 100 g
Formula Weight 293.45
Percent Purity ≥98%
Chemical Name or Material N,N'-Dicyclohexylmorpholine-4-carboxamidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.