missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylurea, TRC

Product Code. 30137111
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137111 50 g 50g
30135564 250 g 250g
30005436 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137111 Supplier Toronto Research Chemicals Supplier No. TRCD43936550G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2387-23-7
Molecular Formula C13 H24 N2 O
Molecular Weight (g/mol) 224.34
Synonym 1,3-Dicyclohexylurea, Lomustine Imp. C (EP), Urea, N,N'-dicyclohexyl-, Urea, 1,3-dicyclohexyl- (6CI,7CI,8CI), N,N'-Dicyclohexylurea, 1,3-Dicyclohexylurea, DCU, Dicyclohexylcarbodiamide, NSC 17013, NSC 30023
IUPAC Name 1,3-dicyclohexylurea
SMILES O=C(NC1CCCCC1)NC2CCCCC2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H24N2O/c16-13(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h11-12H,1-10H2,(H2,14,15,16)
Formula Weight 224.1889
Chemical Name or Material 1,3-Dicyclohexylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.