missing translation for 'onlineSavingsMsg'
Learn More

N,N-DidemethylN,N-diethyl Benzydamine, TRC

Product Code. 30134522
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134522 500 mg 500mg
30004961 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134522 Supplier Toronto Research Chemicals Supplier No. TRCB275750500MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47448-66-8
Molecular Formula C21 H27 N3 O
Molecular Weight (g/mol) 337.46
Synonym 1-Benzyl-3-[3-(diethylamino)propoxy]-1H-indazole, N,N-Diethyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine Hydrochloride;
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-diethylpropan-1-amine
SMILES CCN(CC)CCCOc1nn(Cc2ccccc2)c3ccccc13

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H27N3O/c1-3-23(4-2)15-10-16-25-21-19-13-8-9-14-20(19)24(22-21)17-18-11-6-5-7-12-18/h5-9,11-14H,3-4,10,15-17H2,1-2H3
Formula Weight 337.2154
Percent Purity >95%
Chemical Name or Material 1-Benzyl-3-(3-Diethylamino-Propoxy)-1H-Indazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.