missing translation for 'onlineSavingsMsg'
Learn More

N,N-Didesmethyl Diltiazem Hydrochloride, TRC

Product Code. 30372588
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30372588 10 mg
30380405 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30372588 Supplier Toronto Research Chemicals Supplier No. TRCD44105010MG
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116050-35-2
Molecular Formula C20H23ClN2O4S
Molecular Weight (g/mol) 422.93
Synonym (2S-cis)-3-(Acetyloxy)-5-(2-aminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Monohydrochloride, (2S,3S)-N,N-Didemethyldiltiazem Hydrochloride
SMILES CC(=O)O[C@@H]1[C@@H](SC2=CC=CC=C2N(C1=O)CCN)C3=CC=C(C=C3)OC.Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H22N2O4S.ClH/c1-13(23)26-18-19(14-7-9-15(25-2)10-8-14)27-17-6-4-3-5-16(17)22(12-11-21)20(18)24;/h3-10,18-19H,11-12,21H2,1-2H3;1H/t18-,19+;/m1./s1
Formula Weight 422.1067
Chemical Name or Material N,N-Didesmethyl Diltiazem Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.