missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine, 90%

Product Code. 11358386
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358386 100 g 100g
11338386 5 g 5g
11348386 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11358386 Supplier Thermo Scientific Alfa Aesar Supplier No. L17330.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 357-83-5 | C6H11ClF3N | 189.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357-83-5
Molecular Formula C6H11ClF3N
Molecular Weight (g/mol) 189.606
MDL Number MFCD00054672
InChI Key BDZHKUAKSMWSAJ-UHFFFAOYSA-N
Synonym 2-chloro-1,1,2-trifluoroethyl diethylamine, n,n-diethyl 2-chloro-1,1,2-trifluoroethyl amine, n-2-chloro-1,1,2-trifluoroethyl diethylamine, yarovenko's reagent, acmc-20appo, 2-chloro-1,1,2-trifluorotriethylamine, 1,1,2-trifluoro-2-chloroethyldiethylamine, 2-chloro-1,1,2-trifluoroethyldiethylamine, ethanamine, 2-chloro-n,n-diethyltrifluoro, 1-diethylamino-1,1,2trifluoro-2-chloroethane
PubChem CID 136184
IUPAC Name 2-chloro-N,N-diethyl-1,1,2-trifluoroethanamine
SMILES CCN(CC)C(C(F)Cl)(F)F

Specifications

Density 1.19
Boiling Point 38°C (10 mmHg)
Linear Formula ClFCHCF2N(CH2CH3)2
Quantity 100 g
UN Number UN3267
Beilstein 1098675
Sensitivity Moisture sensitive
Formula Weight 189.61
Percent Purity 90%
Chemical Name or Material N,N-Diethyl(2-chloro-1,1,2-trifluoroethyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.