missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-3-methyl-d3-benzamide-2,4,5,6-d4, TRC
Specifica
Specifica
| Chemical Name or Material | DEET D7 (methyl D3 phenyl D4) |
| CAS | 1219799-37-7 |
| Molecular Formula | C12 2H7 H10 N O |
| Quantity | 2 mg |
| Synonym | N,N-Diethyl-2,4,5,6-tetradeuterio-3-trideuteriomethylbenzamide, DEET D7 (methyl D3 phenyl D4), DEET D7 (methyl D3 benzeneamide D4), Diethyltoluamide-d7 |
| InChI Formula | InChI=1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-7-10(3)9-11/h6-9H,4-5H2,1-3H3/i3D3,6D,7D,8D,9D |
| SMILES | [2H]c1c([2H])c(C(=O)N(CC)CC)c([2H])c(c1[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-N,N-diethyl-5-(trideuteriomethyl)benzamide |
| Molecular Weight (g/mol) | 198.31 |
| Formula Weight | 198.175 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?