missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine oxalate, 96%

Product Code. 10769901 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10769901

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62637-92-7 | 0·5 C2H2O4 | 209.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-92-7
Molecular Formula 0·5 C2H2O4
Molecular Weight (g/mol) 209.27
MDL Number MFCD00070242

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 145.0°C to 150.0°C
Color Brown to Gray or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 10 g
Formula Weight 209.27
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Diethyl-p-phenylenediamine oxalate, 96%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Diethyl-p-phenylenediamine oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.