missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylaniline, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N,N-Diethylaniline |
| CAS | 91-66-7 |
| Molecular Formula | C10 H15 N |
| Quantity | 10 g |
| Synonym | Aniline, N,N-diethyl- (8CI), N,N-Diethylbenzenamine, (Diethylamino)benzene, DEA, Diethylaniline, Diethylphenylamine, N,N-Diethylaminobenzene, N,N-Diethylaniline, N-Phenyldiethylamine, NL 64-10P, NSC 7205 |
| InChI Formula | InChI=1S/C10H15N/c1-3-11(4-2)10-8-6-5-7-9-10/h5-9H,3-4H2,1-2H3 |
| SMILES | CCN(CC)c1ccccc1 |
| IUPAC Name | N,N-diethylaniline |
| Molecular Weight (g/mol) | 149.23 |
| Formula Weight | 149.1204 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction