missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethanolamine(2-Diethylaminoethanol), TRC

Product Code. 30380321
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380321 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30380321 Supplier Toronto Research Chemicals Supplier No. TRCD44377010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(Diethylamino)ethanol
CAS 100-37-8
Molecular Formula C6 H15 N O
Quantity 10 g
Synonym 2-(Diethylamino)ethanol, (2-Hydroxyethyl)diethylamine, (Diethylamino)ethanol, 2-(Diethylamino)ethyl alcohol, 2-(N,N-Diethyl)ethanolamine, 2-(N,N-Diethylamino)ethanol, 2-Hydroxytriethylamine, A 22, A 22 (amine), Amino Alcohol 2A, DEAE, DEEA, Diethyl(2-hydroxyethyl)amine, Diethyl(β-hydroxyethyl)amine, Diethylethanolamine, Diethylmonoethanolamine, MKS, N,N-Diethyl(2-hydroxyethyl)amine, N,N-Diethyl-2-aminoethanol, N,N-Diethylethanolamine, N,N-Diethylmonoethanolamine, N-(2-Hydroxyethyl)diethylamine, NSC 8759, Pennad 150, β-(Diethylamino)ethanol
InChI Formula InChI=1S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3
SMILES CCN(CC)CCO
IUPAC Name 2-(diethylamino)ethanol
Molecular Weight (g/mol) 117.19
Formula Weight 117.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.