missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethanolamine, 99%, Thermo Scientific Chemicals

Product Code. 13458308 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13458308 10 L Plastic drum 10 L
10050880 1 L Glass Bottle 1 L
10236523 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 13458308 Supplier Thermo Scientific Acros Supplier No. 114330100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-37-8
MDL Number MFCD00002850
InChI Key BFSVOASYOCHEOV-UHFFFAOYSA-N
Synonym 2-diethylamino ethanol, n,n-diethylethanolamine, diethylaminoethanol, deae, diethylethanolamine, diethylamino ethanol, ethanol, 2-diethylamino, n,n-diethyl-2-aminoethanol, 2-hydroxyethyl diethylamine, diethyl 2-hydroxyethyl amine
PubChem CID 7497
ChEBI CHEBI:52153
SMILES CCN(CC)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Color White to Yellow
Density 0.8800g/mL
Boiling Point 161.0°C
Flash Point 51.5°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic drum
Molecular Formula C6H15NO
Linear Formula HOCH2CH2N(C2H5)2
Refractive Index 1.4400 to 1.4420
Quantity 10 L
Specific Gravity 0.88
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and benzene
IUPAC Name 2-(diethylamino)ethanol
Molecular Weight (g/mol) 117.192
Formula Weight 117.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Diethylethanolamine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.