missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide, 99%

Product Code. 11409556
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
50 g
250 g
missing translation for 'unitSize':
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
11409556 25 g 25g
11424230 10 g 10g
11434230 50 g 50g
11444230 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11409556 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A19292.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Alfa Aesar N,N'-Diisopropylcarbodiimide, 99%, Quantity: 25g, Boiling Point: 145 deg.C to 148 deg.C, Molecular Weight: 126.203, Percent Purity: 99%, Beilstein: 878281, CAS: 693-13-0, ChEBI: CHEBI:53092, Density: 0.81, DOT Information: Hazard Class: 6.1; Packaging Group: I, EINECS Number: 211-743-7 | CAS: 693-13-0 | C7H14N2 | 126.203 g/mol

N,N'-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diisopropylcarbodiimide is used as a reagent in synthetic organic chemistry. It serves as a chemical intermediate and as a stabilizer for Sarin (chemical weapon). It is also used in the synthesis of peptide and nucleic acid. Further, it is used as an antineoplastic and involved in the treatment of malignant melanoma and sarcomas. In addition to this, it is used in the synthesis of acid anhydride, aldehyde, ketone and isocyanate.

Solubility
Soluble in chloroform, methylene chloride, acetonitrile, dioxane, dimethylformamide andtetrahydrofuran.

Notes
Moisture sensitive. Store in a cool place Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-13-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD00065689
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
PubChem CID 12734
ChEBI CHEBI:53092
IUPAC Name N,N'-di(propan-2-yl)methanediimine
SMILES CC(C)N=C=NC(C)C

Specifications

Density 0.81
Boiling Point 145°C to 148°C
Flash Point 33°C (91°F)
Odor Stench
Refractive Index 1.433
Quantity 25 g
UN Number UN2929
Beilstein 878281
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform,methylene chloride,acetonitrile,dioxane,dimethylformamide andtetrahydrofuran.
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material N,N'-Diisopropylcarbodiimide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N'-Diisopropylcarbodiimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diisopropylcarbodiimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.