missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine, Biotech Grade, 99.5%,Solstice

Product Code. 15645690
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
Packaging:
Glass Bottle
Unit Size:
100mL
2 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15645690 2 L Glass Bottle 2 L
15635690 100 mL Glass Bottle 100mL
15655690 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15645690 Supplier Honeywell-Burdick and Jackson Supplier No. 4962192L
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Biotech Grade, 99.5%

  • Solstice developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Solstice continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

Melting Point -19.99°C
Density 0.742g/mL at 25°C
Boiling Point 128.33°C
Flash Point 12°C (53.6°F)
Odor Strong, amine like
Packaging Glass Bottle
Linear Formula [(CH3)2CH]2NC2H5
Quantity 2 L
UN Number UN3384
Formula Weight 129.24g/mol
Percent Purity 99.50%
Chemical Name or Material N,N-Diisopropylethylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.