missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine, TRC

Product Code. 30040697
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040697 50 g 50g
30005663 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30040697 Supplier Toronto Research Chemicals Supplier No. TRCD49334050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.24
Synonym Triethylamine, 1,1'-dimethyl- (6CI,7CI,8CI), N-Ethyl-N-(1-methylethyl)-2-propanamine, 1,1'-Dimethyltriethylamine, Bis(1-methylethyl)ethylamine, DIEA, DIPEA, Diisopropylethylamine, Ethyl-N,N-diisopropylamine, Ethylbis(propan-2-yl)amine, Ethyldiisopropylamine, Huenig's base, Hunig's base, Hunig's reagent, N,N-Bis(1-methylethyl)-N-ethylamine, N,N-Diisopropylethylamine, N-Ethyl-N,N-diisopropylamine, N-Ethyl-N-isopropylpropan-2-amine, N-Ethyl-N-propan-2-ylpropan-2-amine, N-Ethyldiisopropylamine, NSC 147491, 1,1'-Dimethyltriethylamine, Bis(1-methylethyl)ethylamine, DIEA, DIPEA, Diisopropylethylamine, Ethyl-N,N-diisopropylamine, Ethyldiisopropylamine, Huenig's base, Hunig's base, Hunig's reagent, N,N-Bis(1-methylethyl)-N-ethylamine, N,N-Diisopropylethylamine, N-Ethyl-N,N-diisopropylamine, N-Ethyl-N-(1-methylethyl)-2-propanamine, N-Ethyl-N-isopropylpropan-2-amine, N-Ethyl-N-propan-2-ylpropan-2-amine, N-Ethyldiisopropylamine, NSC 147491
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H19N/c1-6-9(7(2)3)8(4)5/h7-8H,6H2,1-5H3
Formula Weight 129.1517
Percent Purity >95%
Chemical Name or Material N,N-Diisopropylethylamine
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.