missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1-dodecylamine N-oxide, 95%

Product Code. 11322009
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322009 5 g 5g
11332009 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322009 Supplier Thermo Scientific Alfa Aesar Supplier No. H26065.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1643-20-5 | C14H31NO | 229.408 g/mol

N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-20-5
Molecular Formula C14H31NO
Molecular Weight (g/mol) 229.408
MDL Number MFCD00002049
InChI Key SYELZBGXAIXKHU-UHFFFAOYSA-N
Synonym lauramine oxide, lauryldimethylamine oxide, dodecyldimethylamine oxide, lauryldimethylamine n-oxide, ldao, ammonyx ao, ammonyx lo, empigen ob, aromox dmcd, conco xal
PubChem CID 15433
IUPAC Name N,N-dimethyldodecan-1-amine oxide
SMILES CCCCCCCCCCCC[N+](C)(C)[O-]

Specifications

Melting Point 120°C to 123°C
Density 0.996
Quantity 5 g
UN Number UN3259
Beilstein 1769927
Sensitivity Very Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 229.4
Percent Purity 95%
Chemical Name or Material N,N-Dimethyl-1-dodecylamine N-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.