missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine sulfate, 98%

Product Code. 11422237
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422237 500 g 500g
11402237 25 g 25g
11412237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422237 Supplier Thermo Scientific Alfa Aesar Supplier No. A14175.36
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 536-47-0 | C8H14N2O4S | 234.27 g/mol

N,N-Dimethyl-p-phenylenediamine sulfate salt is used to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart, in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin, to investigate colonic tissue H2S production using the acetate trapping assay system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-phenylenediamine sulfate salt is used to investigate the role of H2S in cardioprotection induced by ischemic preconditioning in rat heart, in spectrophotometric determination of gold(III) in gold-pharmaceuticals such as sodium aurothimaleate and auranofin, to investigate colonic tissue H2S production using the acetate trapping assay system.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Store in the dark. Incompatible with oxidizing agents and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-47-0
Molecular Formula C8H14N2O4S
Molecular Weight (g/mol) 234.27
MDL Number MFCD00012992
InChI Key GLUKPDKNLKRLHX-UHFFFAOYSA-N
Synonym n,n-dimethyl-1,4-phenylenediamine sulfate, n,n-dimethyl-p-phenylenediamine sulfate, n1,n1-dimethylbenzene-1,4-diamine sulfate, 1,4-benzenediamine, n,n-dimethyl-, sulfate, unii-x997ivp3ja, 4-amino-n,n-dimethylaniline sulphate, x997ivp3ja, 1,4-benzenediamine, n,n-dimethyl-, sulfate 1:1, dimethyl-p-phenylenediamine; sulfuric acid, 1,4-benzenediamine, n1,n1-dimethyl-, sulfate 1:?
PubChem CID 80351
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine;sulfuric acid
SMILES OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 200°C to 205°C (decomposition)
Odor Odorless
Quantity 500 g
UN Number UN2811
Beilstein 4612278
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 234.28
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.