missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine sulfate, 99%

Product Code. 10062310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10062310 25 g Glass bottle 25g
10164150 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062310 Supplier Thermo Scientific Acros Supplier No. 181430250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 536-47-0 | C8H14N2O4S | 234.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-47-0
Molecular Formula C8H14N2O4S
Molecular Weight (g/mol) 234.27
MDL Number MFCD00012992
InChI Key GLUKPDKNLKRLHX-UHFFFAOYSA-N
Synonym n,n-dimethyl-1,4-phenylenediamine sulfate, n,n-dimethyl-p-phenylenediamine sulfate, n1,n1-dimethylbenzene-1,4-diamine sulfate, 1,4-benzenediamine, n,n-dimethyl-, sulfate, unii-x997ivp3ja, 4-amino-n,n-dimethylaniline sulphate, x997ivp3ja, 1,4-benzenediamine, n,n-dimethyl-, sulfate 1:1, dimethyl-p-phenylenediamine; sulfuric acid, 1,4-benzenediamine, n1,n1-dimethyl-, sulfate 1:?
PubChem CID 80351
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine;sulfuric acid
SMILES OS(O)(=O)=O.CN(C)C1=CC=C(N)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 234°C to 236°C
Color Beige-Gray to Orange or Brown
Boiling Point 495°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2NC6H4NH2·H2SO4
Quantity 25 g
Beilstein 13, IV, 106
Formula Weight 234.28
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N, N-Dimethyl-p-phenylenediamine sulfate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Dimethyl-p-phenylenediamine sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P405-Store locked up.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.