missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-p-phenylenediamine, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl-p-phenylendiamine |
| CAS | 99-98-9 |
| Molecular Formula | C8 H12 N2 |
| Quantity | 100 g |
| Synonym | 1,4-Benzenediamine, N,N-dimethyl- (9CI), p-Phenylenediamine, N,N-dimethyl- (8CI), N1,N1-Dimethyl-1,4-benzenediamine, (4-Aminophenyl)dimethylamine, 1-Amino-4-(dimethylamino)benzene, 4-(Dimethylamino)aniline, 4-(Dimethylamino)benzenamine, 4-(N,N-Dimethylamino)aniline, 4-(N,N-Dimethylamino)phenylamine, 4-Amino-N,N-dimethylaniline, C.I. 76075, DMPD, Dimethyl-p-phenylenediamine, N,N-Dimethyl-1,4-benzenediamine, N,N-Dimethyl-1,4-diaminobenzene, N,N-Dimethyl-1,4-phenylenediamine, N,N-Dimethyl-p-benzenediamine, N,N-Dimethyl-p-phenylenediamine, N-(4-Aminophenyl)-N,N-dimethylamine, NSC 1493, [4-(Dimethylamino)phenyl]amine, p-(Dimethylamino)aniline, p-(Dimethylamino)phenylamine, p-Amino-N,N-dimethylaniline, p-Aminodimethylaniline |
| InChI Formula | InChI=1S/C8H12N2/c1-10(2)8-5-3-7(9)4-6-8/h3-6H,9H2,1-2H3 |
| SMILES | CN(C)c1ccc(N)cc1 |
| IUPAC Name | 4-N,4-N-dimethylbenzene-1,4-diamine |
| Molecular Weight (g/mol) | 136.19 |
| Formula Weight | 136.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction