missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 99+%, extra pure

Product Code. 10502334 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Plastic Drum
Unit Size:
1 L
2.5 L
25 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10502334 25 L Metal Drum 25 L
10154440 1 L Glass Bottle 1 L
10256190 2.5 L Glass Bottle 2.5 L
10047352 5 L Plastic Drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10502334 Supplier Thermo Scientific Acros Supplier No. 181070250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20.0°C
CAS Min % 99.0
CAS Max % 100.0
Density 0.9370g/mL
Quantity 25 L
Flash Point 70°C
Infrared Spectrum Authentic
Packaging Metal Drum
Physical Form Liquid
Refractive Index 1.4370 to 1.4390
Residue after Evaporation 0.01% max.
Assay Percent Range 99+%
Specific Gravity 0.937
Linear Formula CH3CON(CH3)2
Vapor Pressure Vapor Pressure: 1.7mbar at 25°C
Viscosity 1.02 mPa.s (20°C)
Water 0.1% max.
Beilstein 04, 59
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Boiling Point 164.0°C to 166.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Formula Weight 87.12
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.