missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, 99+%, for spectroscopy

Product Code. 10762591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762591 250 mL Glass Bottle 250mL
10012220 1 L Glass Bottle 1 L
10522934 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). all customers must complete a declaration before delivery.
Product Code. 10762591 Supplier Thermo Scientific Acros Supplier No. 167782500
  • ONLINE EXCLUSIVE PRICE
    54.90 €
    50.50 € / 250mL
    Save 4.40 €
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). all customers must complete a declaration before delivery.

CAS: 127-19-5 | C4H9NO | 87.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20.0°C
Absorbance 0.01 max. at 360nm, 0.03 max. at 320nm, 0.05 max. at 310nm, 0.15 max. at 290nm, 0.3 max. at 280nm, 1 max. at 270nm
CAS Min % 99.0
CAS Max % 100.0
Density 0.9370g/mL
Quantity 250 mL
Flash Point 70°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4370 to 1.4390
Residue after Evaporation 0.0005% max.
Assay Percent Range 99+%
Specific Gravity 0.937
Linear Formula CH3CON(CH3)2
Viscosity 1.02 mPa.s (20°C)
Water 0.05% max.
Beilstein 04, 59
Fieser 01,270; 02,144; 04,165
Merck Index 15, 3248
Boiling Point 164.0°C to 166.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone, diethyl ether, benzene,, chloroform, ethanol and toluene
Formula Weight 87.12
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material N, N-Dimethylacetamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.