missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, BAKER ANALYZED™, J.T. Baker™

Product Code. 15578654 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15578654 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578654 Supplier JT Baker Supplier No. 9755.1000
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For headspace gas chromatography

Specifications

CAS 127-19-5
Acidity max. 0.0005 meq/g
Quantity 1 L
Residue after Evaporation max. 0.0005%
Assay Percent Range min. 99.9%
Molecular Formula C4H9NO
MDL Number MFCD00008686
UN Number 2810
Water max. 0.05%
Synonym Acetic acid dimethylamide , Dimethyl Acetamide
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES CN(C)C(C)=O
IUPAC Name N,N-dimethylacetamide
Molecular Weight (g/mol) 87.12
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1LT N,N-DIMETHYLACETAMIDE BAKER ANALYZED GC-HS Reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Dimethylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.