missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, Certified AR for Analysis, Fisher Chemical™

Product Code. 10142140 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Unit Size:
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142140 500 mL Amber glass bottle 500mL
10457502 2.5 L Amber glass bottle 2.5 L
10510341 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10142140 Supplier Fisher Chemical Supplier No. D/3411/08
  • ONLINE EXCLUSIVE PRICE
    64.05 €
    58.20 € / 500mL
    Save 5.85 €
    9% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Melting Point -20°C
Color Colorless
pH 4
Boiling Point 164°C
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 87.12g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.