missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide dimethyl acetal, tech. 90%, stab.

Product Code. 11301367
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301367 5 g 5g
11311367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301367 Supplier Thermo Scientific Alfa Aesar Supplier No. L01642.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 18871-66-4 | C6H15NO2 | 133.191 g/mol

N,N-Dimethylacetamide dimethyl acetal is used as a reagent for the synthesis of amides, diacylamines, and heterocycles, one carbon inserting synthon in preparation of pyrimido[1,2-a][1,3,5]triazin-6-ones. It is used as intermediate in building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylacetamide dimethyl acetal is used as a reagent for the synthesis of amides, diacylamines, and heterocycles, one carbon inserting synthon in preparation of pyrimido[1,2-a][1,3,5]triazin-6-ones. It is used as intermediate in building block.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 2 - 8°C. Incompatible with strong oxidizing agents, Acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18871-66-4
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.191
MDL Number MFCD00008476
InChI Key FBZVZUSVGKOWHG-UHFFFAOYSA-N
Synonym n,n-dimethylacetamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylethylamine, 1,1-dimethoxyethyl dimethylamine, 1,1-dimethoxyethyl dimethyl amine, 1,1-dimethoxy-ethyl-dimethyl-amine, ethanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylacetamide dimethylacetal, dimethylacetamide dimethyl acetal, n,n-dimethylacetamide dimethyl acetal, stabilized with methanol, zlchem 332
PubChem CID 87835
IUPAC Name 1,1-dimethoxy-N,N-dimethylethanamine
SMILES CC(N(C)C)(OC)OC

Specifications

Density 0.911
Boiling Point 118°C to 120°C
Flash Point 8°C (46°F)
Refractive Index 1.412
Quantity 5 g
UN Number UN1993
Beilstein 1420865
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 133.19
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N-Dimethylacetamide dimethyl acetal, stabilized with 5 to 10% methanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.