missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, GC-Headspace tested, ≥99.9%, Solstice

Product Code. 15605560
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15605560 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15605560 Supplier Honeywell-Chromasolv Supplier No. 449011L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, GC-Headspace tested,≥99.9%

Solstice Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests
TRUSTED_SUSTAINABILITY

Specifications

CAS 127-19-5
Density 0.937g/mL at 25°C
Quantity 1 L
Explosion Limit 1.8%, 100°F, 11.5%, 160°F
Flash Point 70°C (158°F)
Ignition Point 914°F
Molecular Formula C4H9NO
Linear Formula CH3CON(CH3)2
Vapor Pressure 4 mmHg (38°C), 2 mmHg (25°C)
MDL Number MFCD00008686
UN Number NA1993
Beilstein 1737614
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES CN(C)C(C)=O
IUPAC Name N,N-dimethylacetamide
Molecular Weight (g/mol) 87.12
PubChem CID 31374
ChEBI CHEBI:84254
Formula Weight 87.12g/mol
Percent Purity ≥99.9% (GC)
Grade CHROMASOLV
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,N-dimethylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.