missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, Puriss. p.a., ≥99.5% (GC), Honeywell Riedel-de Haen™

Product Code. 16625853
Click to view available options
:
50 L
This item is not returnable. View return policy

Product Code. 16625853

Brand: Honeywell Riedel-de Haën 3884050L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 127-19-5
CAS Max % 1
Melting Point -20°C
Color Colorless to Yellow
Density 0.94 g/cm3 (20°C)
pH 4
Boiling Point 164°C to 166°C (1013 hPa)
Flash Point 64°C
Molecular Formula C4H9NO
Linear Formula CH3CON(CH3)2
MDL Number MFCD00008686
Quantity 50 L
Beilstein 1737614
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES CN(C)C(C)=O
IUPAC Name N,N-dimethylacetamide
Molecular Weight (g/mol) 87.12
Grade ACS Grade Plus
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.