missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylallylamine, 98%

Product Code. 10371341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10371341 25 g Glass bottle 25g
10272622 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10371341 Supplier Thermo Scientific Acros Supplier No. 407980250
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2155-94-4 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2155-94-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
MDL Number MFCD00038234
InChI Key GBCKRQRXNXQQPW-UHFFFAOYSA-N
Synonym n,n-dimethylallylamine, allyldimethylamine, dimethylallylamine, 2-propen-1-amine, n,n-dimethyl, 1-dimethylamino-2-propene, n-allyl-n,n-dimethylamine, n-allyldimethylamine, dimethyl prop-2-en-1-yl amine, allylamine, n,n-dimethyl, ch2=chch2n ch3 2
PubChem CID 75082
IUPAC Name N,N-dimethylprop-2-en-1-amine
SMILES CN(C)CC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7410g/mL
Boiling Point 62°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHCH2N(CH3)2
Refractive Index 1.4000 to 1.402
Quantity 25 g
Specific Gravity 0.741
Solubility Information Solubility in water: soluble
Formula Weight 85.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylallylamine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.