missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, TRC

Product Code. 30372663
Change view
Click to view available options
Quantity:
50 mL
250 mL
500 mL
Unit Size:
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30372663 50 mL -
30372269 500 mL 500mL
30376386 250 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372663 Supplier Toronto Research Chemicals Supplier No. TRCD46410050ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8 H11 N
Molecular Weight (g/mol) 121.18
Synonym Aniline, N,N-dimethyl- (8CI), N,N-Dimethylbenzenamine, (Dimethylamino)benzene, Accelerator NL 63-100, DMA, Dimethylaniline, Dimethylphenylamine, EP 210, N,N-Dimethylaminobenzene, N,N-Dimethylaniline, N,N-Dimethylphenylamine, NL 63-10P, NSC 7195, Versneller NL 63/10
IUPAC Name N,N-dimethylaniline
SMILES CN(C)c1ccccc1

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3
Formula Weight 121.0891
Chemical Name or Material N,N-Dimethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.