missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzylamine(Benzyldimethylamine), TRC

Product Code. 30336111
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336111 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336111 Supplier Toronto Research Chemicals Supplier No. TRCB27620050ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylbenzylamine(Benzyldimethylamine)
CAS 103-83-3
Molecular Formula C9H13N
Quantity 50 mL
Synonym N,N-Dimethylbenzenemethanamine, 4-[(Dimethylamino)methyl]styrene, Actiron NX 91, Ancamine BDMA, Araldite Accelerator 062, Araldite DY 062, BDMA, Benzyl-N,N-dimethylamine, DY 062, DY 62, Dabco BDMA, Desmorapid DB, Dimethylbenzylamine, Kaolizer 20, N,N-Dimethyl-N-benzylamine, N,N-Dimethylbenzenemethanamine, N-(Phenylmethyl)dimethylamine, N-Benzyl-N,N-dimethylamine, N-Benzyldimethylamine, NSC 5342, Sumicure BD, Y 27, [(Dimethylamino)methyl]benzene
InChI Formula InChI=1S/C9H13N/c1-10(2)8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3
SMILES CN(C)Cc1ccccc1
IUPAC Name N,N-dimethyl-1-phenylmethanamine
Molecular Weight (g/mol) 135.21
Formula Weight 135.1048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.