missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylcarbamothioic chloride, Thermo Scientific™
Specifications
Specifications
| CAS | 16420-13-6 |
| Molecular Formula | C3H6ClNS |
| MDL Number | MFCD00004919 |
| Quantity | 25 g |
| Synonym | dimethylthiocarbamoyl chloride, carbamothioic chloride, dimethyl, chloromethanethioyl dimethylamine, n,n-dimethylthiocarbamoyl chloride, n,n-dimethylcarbamothioic chloride, dimethylthiocarbamoylchloride, carbamoyl chloride, dimethylthio, carbamothioic chloride, n,n-dimethyl, dimethylthiocarbamyl chloride, dimethylthiocarbamic acid chloride |
| InChI Key | PHWISQNXPLXQRU-UHFFFAOYSA-N |
| SMILES | CN(C)C(Cl)=S |
| IUPAC Name | N,N-dimethylcarbamothioyl chloride |
| Molecular Weight (g/mol) | 123.60 |
| PubChem CID | 27871 |
| Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?