missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylcyclohexylamine, 99%

Product Code. 10508512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10508512 500 g Glass bottle 500g
10095403 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10508512 Supplier Thermo Scientific Acros Supplier No. 408255000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-94-2 | C8H17N | 127.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-94-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00003844
InChI Key SVYKKECYCPFKGB-UHFFFAOYSA-N
Synonym n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl
PubChem CID 7415
ChEBI CHEBI:59022
IUPAC Name N,N-dimethylcyclohexanamine
SMILES CN(C)C1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -60.0°C
Density 0.8400g/mL
Boiling Point 160.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4525 to 1.4545
Quantity 500 g
Specific Gravity 0.84
Solubility Information Solubility in water: 13g/L (20°C)
Viscosity 1.16 mPa.s (25°C)
Formula Weight 127.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylcyclohexylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylcyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.