missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldodecylamine, 95%, pract.

Product Code. 10209500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209500 250 g Glass bottle 250g
10269400 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10209500 Supplier Thermo Scientific Acros Supplier No. 408292500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-18-5 | C14H31N | 213.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-18-5
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.41
MDL Number MFCD00008970
InChI Key YWFWDNVOPHGWMX-UHFFFAOYSA-N
Synonym n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
PubChem CID 8168
IUPAC Name N,N-dimethyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -20.0°C
Density 0.7870g/mL
Boiling Point 271.0°C
Flash Point 118°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Dodecyl) (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)11N(CH3)2
Refractive Index 1.4365 to 1.4385
Quantity 250 g
Specific Gravity 0.787
Solubility Information Solubility in water: negligible
Viscosity 1.8 mPa.s (100°C)
Formula Weight 213.41
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyldodecylamine, pract., 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyldodecylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.