missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethanolamine, 99%

Código de producto. 10132480 Tienda Thermo Scientific Chemicals Productos
Cambiar vista
Click to view available options
Quantity:
250 mL
1 L
5 L
25 L
Packaging:
Glass bottle
Metal Drum
Plastic drum
Tamaño de la unidad:
1 L
25 L
250mL
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging unitSize
10132480 250 mL Glass bottle 250mL
10112480 1 L Glass bottle 1 L
10358793 5 L Plastic drum 5 L
10594683 25 L Metal Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 10132480 Proveedor Thermo Scientific Acros N.º de proveedor 116182500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 108-01-0 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 108-01-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00002846
InChI Key UEEJHVSXFDXPFK-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanol, n,n-dimethylethanolamine, deanol, dimethylaminoethanol, dimethylethanolamine, norcholine, dmae, dmea, bimanol, liparon
PubChem CID 7902
ChEBI CHEBI:271436
IUPAC Name 2-(dimethylamino)ethanol
SMILES CN(C)CCO

Especificaciones

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.8880g/mL
Boiling Point 139.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH2N(CH3)2
Refractive Index 1.4290 to 1.4300
Quantity 250 mL
Beilstein 04, 276
Merck Index 15, 2845
Specific Gravity 0.888
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and ether
Viscosity 3.8 mPa.s (20°C)
Formula Weight 89.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylethanolamine
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • N,N-Dimethylethanolamine
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.