missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylamine, 99%

Product Code. 10360844 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
This item is not returnable. View return policy

Product Code. 10360844

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-56-1 | C4H11N | 73.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-56-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.13
MDL Number MFCD00009039
InChI Key DAZXVJBJRMWXJP-UHFFFAOYSA-N
Synonym n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl
PubChem CID 11723
IUPAC Name N,N-dimethylethanamine
SMILES CCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -140°C
Color Colorless to Yellow
Density 0.6750g/mL
Boiling Point 36°C
Flash Point -36°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C2H5N(CH3)2
Refractive Index 1.3715 to 1.3735
Quantity 1 L
Beilstein 04, 94
Specific Gravity 0.675
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most common organic solvents
Formula Weight 73.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylethylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.