missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylethylenediamine, 95%

Product Code. 11331547
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331547 5 g 5g
11341547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331547 Supplier Thermo Scientific Alfa Aesar Supplier No. L02204.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 110-70-3 | C4H12N2 | 88.154 g/mol

N,N'-Dimethylethylenediamine is used in DNA binding effect. It is used to enhance the adsorption of carbon dioxide. It acts as a ligand and form coordination complex such as dinitrato(N,N'-dimethyl-1,2-ethanediamine)copper(II) and dichloro(1,4-bis-(diphenyl phosphino)butane)-(1,2-ethylenediamine)ruthenium(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dimethylethylenediamine is used in DNA binding effect. It is used to enhance the adsorption of carbon dioxide. It acts as a ligand and form coordination complex such as dinitrato(N,N′-dimethyl-1,2-ethanediamine)copper(II) and dichloro(1,4-bis-(diphenyl phosphino)butane)-(1,2-ethylenediamine)ruthenium(II).

Solubility
Miscible with chloroform and dichloromethane.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-70-3
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008290
InChI Key KVKFRMCSXWQSNT-UHFFFAOYSA-N
Synonym n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
PubChem CID 8070
IUPAC Name N,N'-dimethylethane-1,2-diamine
SMILES CNCCNC

Specifications

Density 0.818
Boiling Point 118°C to 119°C
Flash Point 28°C (82°F)
Odor Amine-like
Linear Formula CH3NHCH2CH2NHCH3
Refractive Index 1.43
Quantity 5 g
UN Number UN2734
Beilstein 878142
Sensitivity Air Sensitive
Solubility Information Miscible with chloroform and dichloromethane.
Formula Weight 88.15
Percent Purity 95%
Chemical Name or Material N,N'-Dimethylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.