missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylenediamine, 97%

Product Code. 11459616
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459616 50 g 50g
11478770 100 g 100g
11469616 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459616 Supplier Thermo Scientific Alfa Aesar Supplier No. B20182.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alfa Aesar N,N-Dimethylethylenediamine, 97%, Quantity: 250g, Boiling Point: 107 deg.C, Melting Point: <-70 deg.C, Molecular Weight: 88.154, Percent Purity: 97%, Beilstein: 605279, CAS: 108-00-9, Density: 0.81, DOT Information: Hazard Class: 8; Packaging Group: I, EINECS Number: 203-541-2 | CAS: 108-00-9 | C4H12N2 | 88.154 g/mol

It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as an intermediate in the synthesis of Cefotiam. It is also used as a catalyst in the synthesis of allylic nitro compounds.

Solubility
Fully miscible in water.

Notes
Air sensitive & hygroscopic. Store away from oxidizing agents, air and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-00-9
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008175
InChI Key DILRJUIACXKSQE-UHFFFAOYSA-N
Synonym n,n-dimethylethylenediamine, 2-dimethylaminoethylamine, 2-aminoethyl dimethylamine, 1,2-ethanediamine, n,n-dimethyl, 2-dimethylamino ethylamine, n,n-dimethyl-1,2-ethanediamine, n1,n1-dimethylethane-1,2-diamine, n,n-dimethyl-1,2-ethylenediamine, 2-aminoethyldimethylamine, n,n-dimethylethanediamine
PubChem CID 66053
IUPAC Name N',N'-dimethylethane-1,2-diamine
SMILES CN(C)CCN

Specifications

Melting Point <-70°C
Density 0.81
Boiling Point 107°C
Flash Point 12°C (53°F)
Linear Formula (CH3)2NCH2CH2NH2
Refractive Index 1.427
Quantity 50 g
UN Number UN2733
Beilstein 605279
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 88.15
Percent Purity 97%
Chemical Name or Material N,N-Dimethylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.