missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylethylenediamine, tech., 85%

Product Code. 10214240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10214240 25 g Glass bottle 25g
10539420 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214240 Supplier Thermo Scientific Acros Supplier No. 116190250
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 110-70-3 | C4H12N2 | 88.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-70-3
InChI Key KVKFRMCSXWQSNT-UHFFFAOYSA-N
Synonym n,n'-dimethylethylenediamine, 1,2-bis methylamino ethane, n1,n2-dimethylethane-1,2-diamine, n,n'-dimethyl-1,2-ethanediamine, dimethylethylenediamine, sym-dimethylethylenediamine, 1,2-ethanediamine, n,n'-dimethyl, 2,5-diazahexane, n,n'-dimethyldiaminoethane, n,n'-dimethylethanediamine
PubChem CID 8070
IUPAC Name N,N'-dimethylethane-1,2-diamine
SMILES CNCCNC

Specifications

Color Colorless to Yellow
Density 0.8190g/mL
Boiling Point 119.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H12N2
Linear Formula CH3NHCH2CH2NHCH3
Refractive Index 1.4295
Quantity 25 g
Beilstein 04, 250
Specific Gravity 0.819
Solubility Information Solubility in water: miscible. Other solubilities: soluble in organic solvents
Molecular Weight (g/mol) 88.15
Formula Weight 88.15
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material N,N′-Dimethylethylenediamine, tech., 85%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.