missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, ≥99.8%, ACS Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™

Product Code. 15621460
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Unit Size:
2 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15621460

Brand: Honeywell-Riedel-de Haen 1548142L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Specifications

CAS 68-12-2
Density 0.944g/mL
Quantity 2 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Packaging Glass Bottle
Molecular Formula C3H7NO
Linear Formula HCON(CH3)2
MDL Number MFCD00003284
Vapor Pressure 2.7mmHg (20°C)
UN Number UN2265
Beilstein 605365
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES CN(C)C=O
IUPAC Name N,N-dimethylformamide
Molecular Weight (g/mol) 73.10
PubChem CID 6228
ChEBI CHEBI:17741
Formula Weight 73.09g/mol
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.