missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99+%, extra pure

Product Code. 10590571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
2.5 L
25 L
5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10590571 1 L Glass Bottle 1 L
10112530 2.5 L Glass Bottle 2.5 L
10271233 5 L Plastic Drum 5 L
10733651 25 L Plastic Drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10590571 Supplier Thermo Scientific Acros Supplier No. 116220010
  • ONLINE EXCLUSIVE PRICE
    54.10 €
    50.00 € / 1 L
    Save 4.10 €
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61.0°C
CAS Min % 99.0
CAS Max % 100.0
Decomposition Information >350°C
Density 0.9450g/mL
Quantity 1 L
Flash Point 58°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4290 to 1.4320
Residue after Evaporation 0.005% max.
Assay Percent Range 99+%
Specific Gravity 0.945
Linear Formula HCON(CH3)2
Vapor Pressure Vapor Pressure: 4.9mbar at 20°C
Viscosity 0.8 mPa.s (20°C)
Water 0.2% max. (K.F.)
Beilstein 04, 58
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Boiling Point 153.0°C
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Formula Weight 73.09
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material N,N-Dimethylformamide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: dimethyl formamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.