missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Azine Dihydrochloride, TRC

Product Code. 30375539
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375539 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375539 Supplier Toronto Research Chemicals Supplier No. TRCD4730951G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylformamide Azine Dihydrochloride
CAS 868670-46-6
Molecular Formula C6H16Cl2N4
Quantity 1 g
Synonym 1,2-Bis[(dimethylamino)methylene]hydrazine Dihydrochloride, Dimethylformamide Azine Dihydrochloride, N,N-Dimethyl Formamide Azine Dihydrochloride, N'-[(dimethylamino)methylene]-N,N-dimethyl-methanehydrazonamide Dihydrochloride
InChI Formula InChI=1S/C6H14N4.2ClH/c1-9(2)5-7-8-6-10(3)4;;/h5-6H,1-4H3;2*1H/b7-5+,8-6+;;
SMILES Cl.Cl.CN(C)\C=N\N=C\N(C)C
IUPAC Name N'-[(E)-dimethylaminomethylideneamino]-N,N-dimethylmethanimidamide;dihydrochloride
Molecular Weight (g/mol) 215.12
Formula Weight 214.0752
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.