missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide-d{7}, 99.5% (Isotopic)

Product Code. 11319977
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
5 x 1 g
Unit Size:
10g
1g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319977 1 g 1g
11316079 5 x 1 g 5g
15401807 5 g 5g
15411807 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11319977 Supplier Thermo Scientific Alfa Aesar Supplier No. 042284.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 4472-41-7 | C3H7NO | 80.138 g/mol

It is a common solvent used in NMR spectroscopy. It is also used as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3H7NO
Molecular Weight (g/mol) 80.138
MDL Number MFCD00003286
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym n,n-dimethylformamide-d7, dmf-d7, deuterated dmf, n,n-di 2h3 methyl 2h formamide, dimethyl formamide-d7, dimethylformamide d, heptadeutero-n,n-dimethylformamide, n,n-dimethylformamide-d7 dmf-d7, formamide-1-d, n,n-di methyl-d3, n,n-dimethylformamide-d7 99.5atom%d
PubChem CID 78225
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
SMILES CN(C)C=O

Specifications

Physical Form Liquid
Color Colorless
Density 1.04 g/mL
Flash Point 58°C (136°F)
Odor Amine-like
Packaging Prescored Ampule
Quantity 1 g
Boiling Point 153°C
Linear Formula DCON(CD3)2
UN Number UN2265
Solubility Information Soluble in water.
Formula Weight 80.14
Percent Purity 99.5%
Chemical Name or Material N,N-Dimethylformamide-d7, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.