missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide-d7, TRC

Product Code. 30041210
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041210 5 g 5g
30040455 500 mg 500mg
30041087 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30041210 Supplier Toronto Research Chemicals Supplier No. TRCD4469975G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3 2H7 N O
Molecular Weight (g/mol) 80.14
Synonym DMF-d7, Dimethylformamide-d7, N,N-Dimethylformamide-d7, Perdeuterodimethylformamide
IUPAC Name 1-deuterio-N,N-bis(trideuteriomethyl)formamide
SMILES [2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3/i1D3,2D3,3D
Formula Weight 80.0967
Percent Purity >95%
Chemical Name or Material N,N-Dimethylformamide-d7
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.