missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide Di-tert-butyl Acetal, TRC

Product Code. 30381534
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381534 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381534 Supplier Toronto Research Chemicals Supplier No. TRCD4649952.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethylformamide Di-tert-butyl Acetal
CAS 36805-97-7
Molecular Formula C11H25NO2
Quantity 2.5 g
Synonym 1,1-bis(1,1-Dimethylethoxy)-N,N-dimethylmethanamine, 1,1-Di-tert-butoxytrimethylamine, 1,1-Di-tert-butoxy-N,N-dimethylmethanamine, 1,1-Di-tert-butoxy-N,N-dimethylmethaneamine, DMF Di-tert-butyl Acetal, Di(tert-butoxy)(dimethylamino)methane, Di-tert-butoxymethyldimethylamine, Di-tert-butyl Dimethylformamide Acetal, Dimethylformamide Di-tert-butyl Acetal, N,N-DMF Di-tert-butyl Acetal
InChI Formula InChI=1S/C11H25NO2/c1-10(2,3)13-9(12(7)8)14-11(4,5)6/h9H,1-8H3
SMILES CN(C)C(OC(C)(C)C)OC(C)(C)C
IUPAC Name N,N-dimethyl-1,1-bis[(2-methylpropan-2-yl)oxy]methanamine
Molecular Weight (g/mol) 203.32
Formula Weight 203.1885
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.