missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide dimethyl acetal, 94%

Product Code. 11438276
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438276 500 g 500g
11418276 25 g 25g
11428276 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11438276 Supplier Thermo Scientific Alfa Aesar Supplier No. A15350.36
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol

N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide dimethyl acetal is used as an intermediate in the formation of pyridine derivatives, which exhibits inhibition against PI3 kinase p110alfa enzymes. It is utilized for the derivatization of primary sulfonamides and trifluoroacetic acid. It is also used in the preparation of formamidine derivatives. It is used as a reagent for n-dimethylaminomethylene and methyl esters. Further, it is used to catalyze the coupling of epoxides with carbon dioxide to prepare cyclic carbonates.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4637-24-5
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00008482
InChI Key ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
PubChem CID 78373
ChEBI CHEBI:85061
IUPAC Name (dimethoxymethyl)dimethylamine
SMILES COC(OC)N(C)C

Specifications

Melting Point -85°C
Density 0.897
Boiling Point 102°C to 104°C
Flash Point 7°C (45°F)
Refractive Index 1.3972
Quantity 500 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information Miscible with most organic solvents.
Formula Weight 119.16
Percent Purity 97%
Chemical Name or Material N,N-Dimethylformamide dimethyl acetal
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Dimethylformamide dimethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.