Product Code. 10317390
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
10317390
500 g
Glass bottle
500g
10001900
25 g
Glass bottle
25g
10687322
100 g
Glass bottle
100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code.
10317390
Supplier
Thermo Scientific Acros
Supplier No.
161255000
680.20 EUR valid until 2026-12-31 BEST PRICE on Promo! Use promo code "27993" to get your promotional price.
missing translation for 'validMainframeMsg'
Normativo: Questo prodotto è un intermedio isolato trasportato in conformità con il regolamento (CE) 1907/2006 (REACH). Tutti i clienti devono compilare una dichiarazione prima della consegna.
CAS: 4637-24-5 | C5H13NO2 | 119.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
Identificatori chimici
CAS
4637-24-5
,
149-73-5
,
68-12-2
,
67-56-1
Molecular Formula
C5H13NO2
Molecular Weight (g/mol)
119.16
MDL Number
MFCD00008482
InChI Key
ZSXGLVDWWRXATF-UHFFFAOYSA-N
Synonym
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
PubChem CID
78373
ChEBI
CHEBI:85061
IUPAC Name
(dimethoxymethyl)dimethylamine
SMILES
COC(OC)N(C)C
Specifica
CAS Min %
96.0
CAS Max %
100.0
Color
Colorless
Density
0.8900g/mL
Boiling Point
102.0°C to 104.0°C
Flash Point
7°C
Infrared Spectrum
Authentic
Assay Percent Range
96% min. (GC)
Packaging
Glass bottle
Linear Formula
(CH3 )2 NCH(OCH3 )2
Refractive Index
1.3972
Quantity
500 g
UN Number
1993
Fieser
03,115; 04,184; 05,254; 06,221; 08,191; 09,182; 10,158; 13,120; 05,646; 06,566; 07,355; 09,447; 11,501
Specific Gravity
0.89
Solubility Information
Solubility in water: hydrolysis. Other solubilities: soluble in many organic solvents
Formula Weight
119.16
Percent Purity
ca. 97%
Physical Form
Liquid
Chemical Name or Material
N, N-Dimethylformamide dimethyl acetal, ca 97%
Vedi altri risultati
Mostra meno risultati
Identificativo prodotto
N,N-Dimethylformamide dimethyl acetal
Avvertenza
Categoria di pericolo
Acute toxicity Category 4
Serious eye damage/eye irritation Category 1
Flammable liquid Category 2
Skin sensitiser Category 1
Indicazioni di pericolo
H225-Highly flammable liquid and vapour.
H317-May cause an allergic skin reaction.
H318-Causes serious eye damage.
H332-Harmful if inhaled.
Consigli di prudenza
P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
P280-Wear protective gloves/protective clothing/eye protection/face protection.
P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
MIXTURE LIST-Contain: N,N-Dimethylformamide dimethyl acetal
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy.
Informativa sulla privacy .
Individuate un'opportunità di miglioramento?