missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide dineopentyl acetal, 98%

Product Code. 11387805
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387805 10 g 10g
11397805 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387805 Supplier Thermo Scientific Alfa Aesar Supplier No. L02728.09
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4909-78-8 | C13H30NO2 | 232.39 g/mol

N,N-Dimethylformamide dineopentyl acetal helps in the lactonization of ω-hydroxyacids, giving lactones of up to 16-membered rings. It can be used to convert primary alcohols to alkylating agents for use in the alkylation of thiols. It is used in the esterification of Nα-9-fluorenylmethyloxycarbonylamino acids. It was used in the synthesis of 1,3-dialkyl, benzyl and cyclohexyl barbiturate derivatives. It was used as reagent during the synthesis of L-serine and L-cystine stereospecifically labeled with deuterium at the β-position.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylformamide dineopentyl acetal helps in the lactonization of ω-hydroxyacids, giving lactones of up to 16-membered rings. It can be used to convert primary alcohols to alkylating agents for use in the alkylation of thiols. It is used in the esterification of Nα-9-fluorenylmethyloxycarbonylamino acids. It was used in the synthesis of 1,3-dialkyl, benzyl and cyclohexyl barbiturate derivatives. It was used as reagent during the synthesis of L-serine and L-cystine stereospecifically labeled with deuterium at the β-position.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Incompatible with water and oxidizing agents. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4909-78-8
Molecular Formula C13H30NO2
Molecular Weight (g/mol) 232.39
MDL Number MFCD00008851
InChI Key KEXFRBIOHPDZQM-UHFFFAOYSA-O
Synonym n,n-dimethylformamide dineopentyl acetal, methanamine, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethyl, n,n-dimethyl-1,1-bis neopentyloxy methanamine, dimethylformamide dineopentyl acetal, 1,1-bis 2,2-dimethylpropoxy-n,n-dimethylmethanamine, bis 2,2-dimethylpropoxy methyl dimethylamine, acmc-1al8s, 1,1-bis 2,2-dimethylpropoxy-n,n,n-trimethylamine, 1,1-dineopentyloxytrimethylamine, dimethylformamide dineopentylacetal
PubChem CID 78623
IUPAC Name 1,1-bis(2,2-dimethylpropoxy)-N,N-dimethylmethanamine
SMILES C[NH+](C)C(OCC(C)(C)C)OCC(C)(C)C

Specifications

Density 0.83
Boiling Point 85°C to 87°C (10 mmHg)
Flash Point 63°C (145°F)
Linear Formula (CH3)2NCH[OCH2C(CH3)3]2
Refractive Index 1.4117
Quantity 10 g
Beilstein 741992
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 231.38
Percent Purity 98%
Chemical Name or Material N,N-Dimethylformamide dineopentyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.