missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Product Code. 13104829
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
13104829 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 13104829 Supplier Merck Chemicals Supplier No. 1109831000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7 (at 200g/L 20°C)
Melting Point -61°C
Color Colorless
Density 0.94g/cm3 at 20°C
Quantity 1 L
Flash Point 58°C (136°F)
Ignition Point 410°C (770°F)
Odor Weakly amine-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.43
Vapor Pressure 3.77 hPa (20°C)
UN Number 2265
Boiling Point 153°C
Solubility Information Soluble in water at 20°C
Formula Weight 73.09
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.