missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylformamide, GC-Headspace tested, ≥99.9%, Solstice
Description
Solstice Chromasolv™ GC-headspace solvents:
- developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- very pure solvents with extremely low concentrations of the defined residual solvents
- state-of-the art production and application tests
Spécification
Spécification
| Melting Point | -61°C (literature) |
| Absorbance | max. 0.700 at 270 nm, max. 0.250 at 275 nm, max. 0.070 at 300 nm, max. 0.022 at 320 nm |
| CAS | 68-12-2 |
| CAS Min % | ≥90.0000% |
| CAS Max % | ≤100.0000% |
| Density | 0.944 g/mL(lit.) |
| Quantity | 1 L |
| Flash Point | 57.5°C |
| Packaging | Glass Bottle |
| Molecular Formula | C3H7NO |
| Afficher plus |
Safety and Handling
Identifiant produit
- N,N-Dimethylformamide
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-[In case of inadequate ventilation] wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: N,N-dimethylformamide
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?