missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, puriss. p.a. ACS Reagent, Solstice

Product Code. 15653900
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
180 kg
Unit Size:
1 L
180kg
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15653900 2.5 L 2.5 L
15643900 1 L 1 L
15673900 25 L 25 L
15633900 180 kg 180kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15653900 Supplier Fluka Supplier No. 331202.5L
  • ONLINE EXCLUSIVE PRICE
    193.00 €
    49.53 € / 2.5 L
    Save 143.47 €
    74% Off
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Density 0.944g/mL, 0.948 to 0.950g/mL at 20°C
Quantity 2.5 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20°C)
UN Number 2265
Beilstein 605365
Formula Weight 73.09g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material N,N-Dimethylformamide, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312+H332-Harmful in contact with skin or if inhaled
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.