missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.8% (GC),Honeywell™ Riedel-de Haën™

Product Code. 15683900
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
2.5 L
Pack of 4
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15683900 4 x 2.5 L Glass Bottle Pack of 4
15663900 2.5 L Poly Bottle 2.5 L
15693900 6 x 1 L Glass Bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15683900 Supplier Honeywell-Riedel-de Haen Supplier No. 331204X2.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

pH 7 (at 20°C)
Melting Point -61°C
Density 0.944g/mL, 0.948 to 0.950g/mL at 20°C
Quantity 4 x 2.5 L
Explosion Limit 0.152
Flash Point 58°C (136.4°F)
Ignition Point 833°F
Odor Weak
Packaging Glass Bottle
Physical Form Liquid
Linear Formula HCON(CH3)2
Vapor Pressure 2.7mmHg (20°C)
UN Number UN2265
Beilstein 605365
Boiling Point 152°C to 154°C
Formula Weight 73.09g/mol
Percent Purity ≥99.8% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material N,N-Dimethylformamide, Reag. Ph. Eur.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.